3-Phenoxybenzyl chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
590932-1G 590932-5G
SIAL590932-1GEA 66.4 EUR
SIAL590932-1G SIAL590932-5G
3-Phenoxybenzyl chloride, Sigma-Aldrich®
3-Phenoxybenzyl chloride
Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Boiling Pt: 123 °C (0.3 mmHg)
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00040866
CAS Number: 53874-66-1
EINECS: 258-831-1
UN: 3265
ADR: 8 III