Valerophenone, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
V659-10G V659-100G V659-25G
SIALV659-10GEA 26 EUR
SIALV659-10G SIALV659-100G SIALV659-25G
Valerophenone, Sigma-Aldrich®
Valerophenone
Formula: C₆H₅C(O)(CH₂)₃CH₃
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: –9 °C
Density: 0.98777 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009480
CAS Number: 1009-14-9
EINECS: 213-767-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2584f16b192b481fbaf1d8ab1e77be6109fb67a6&vid=3bf6c22512cdad990d29bc1ffe3db3b535a55970&prid=web&clid=SAPDAM