Propiophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Phenyl ethyl ketone, Ethyl phenyl ketone, 1-Phenyl-1-propanone
M-Clarity™ quality level = MQ200
P51605-100G P51605-500G
SIALP51605-100GEA 36.8 EUR
SIALP51605-100G SIALP51605-500G
Propiophenone, Sigma-Aldrich®
Propiophenone
Formula: C₆H₅C(O)CH₂CH₃
MW: 134.18 g/mol
Boiling Pt: 217
Melting Pt: 18
Density: 1.01 g/cm³ (20 °C)
Flash Pt: 85
Storage Temperature: Ambient
MDL Number: MFCD00009309
CAS Number: 93-55-0
EINECS: 202-257-6
Merck Index: 12,08015
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=796cda47787bc1a939c0285780228746da70cd03&vid=b0290578193d65d6b598ec25da82aff9335f1d45&prid=web&clid=SAPDAM