3-Acetylbenzoic acid, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
253839-1G
253839-5G
SIAL253839-1GEA
25.8
EUR
SIAL253839-1G
SIAL253839-5G
3-Acetylbenzoic acid, Sigma-Aldrich®
3-Acetylbenzoic acid
Formula:
C₉H₈O₃ Boiling Pt: 343 °C (1013 hPa) Melting Pt: 172 °C Density: 1.23 g/cm³ (20 °C) |
MDL Number:
MFCD00045847 CAS Number: 586-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...