4'-Nitroacetophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms:
p-Nitroacetophenone, 1-(4-Nitrophenyl)ethanone
M-Clarity™ quality level = MQ100
N9608-100G
N9608-5G
SIALN9608-100GEA
56.6
EUR
SIALN9608-100G
SIALN9608-5G
4'-Nitroacetophenone, Sigma-Aldrich®
4'-Nitroacetophenone
Formula:
C₈H₇NO₃ Boiling Pt: 275 °C (1013 hPa) Melting Pt: 76…78 °C Flash Pt: 149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007355 CAS Number: 100-19-6 EINECS: 202-827-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...