2-Toluoyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Methylbenzoyl chloride, o-Methylbenzoyl chloride, o-Toluoyl chloride, 2-Methylbenzene-1-carbonyl chloride
M-Clarity™ quality level = MQ100
122017-100G 122017-5G
SIAL122017-100GEA 122 EUR
SIAL122017-100G SIAL122017-5G
2-Toluoyl chloride, Sigma-Aldrich®
2-Toluoyl chloride
Formula: C₈H₇ClO
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1.186 g/cm³ (20 °C)
MDL Number: MFCD00000668
CAS Number: 933-88-0
EINECS: 213-273-8
UN: 3265
ADR: 8,II