2-Fluorotoluene, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Fluoro-2-methylbenzene, o-Fluorotoluene
M-Clarity™ quality level = MQ100
F15323-100G F15323-500G
SIALF15323-100GEA 50 EUR
SIALF15323-100G SIALF15323-500G
2-Fluorotoluene, Sigma-Aldrich®
2-Fluorotoluene
Formula: C₇H₇F
MW: 110.13 g/mol
Boiling Pt: 113
Melting Pt: –62 °C
Density: 1.005 g/cm³ (20 °C)
Flash Pt: 8
Storage Temperature: Ambient
MDL Number: MFCD00000322
CAS Number: 95-52-3
EINECS: 202-428-5
UN: 2388
ADR: 3 II
Merck Index: 13,04207