α,α,α-Trifluoro-2-tolualdehyde
Supplier: Thermo Scientific
Warning
SB01906.250 SB01906.1
MAYBSB01906.250EA 0 EUR
MAYBSB01906.250 MAYBSB01906.1
α,α,α-Trifluoro-2-tolualdehyde
α,α,α-Trifluoro-2-tolualdehyde
Formula: C₈H₅F₃O
Boiling Pt: 28…38 °C (0.4 mmHg)
Melting Pt: –40 °C
Density: 1,322 g/cm³ (20 °C)
MDL Number: MFCD00003337
CAS Number: 447-61-0
EINECS: 207-185-9