(4-Fluorophenyl)acetonitrile ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14290.06
A14290.14
A14290.22
A14290.06EA
25.6
EUR
A14290.06
A14290.14
A14290.22
(4-Fluorophenyl)acetonitrile ≥98%
(4-Fluorophenyl)acetonitrile
Purity: 98+%
CAS No.: 459-22-3
Molecular Formula: C8H6FN
Formula Weight: 135.14
Flash Point: 108°(226°F)
Refractive Index: 1.5002
Boiling Point: 238°
Beilstein Reference: 1907764
UN No.: 3276
MDL No.: MFCD00001917
Formula:
C₈H₆FN Boiling Pt: 238 °C Melting Pt: 86 °C Density: 1.130 Flash Pt: 108 °C (226 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001917 CAS Number: 459-22-3 EINECS: 207-286-8 UN: 3276 ADR: 6.1,III |