α,α,α-Trifluoroacetophenone, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2,2,2-Trifluoroacetophenone, Phenacyl trifluoride, Trifluoroacetylbenzene, 1-Phenyl-2,2,2-trifluoro-1-ethanone
M-Clarity™ quality level = MQ200
107840-100G
107840-25G
107840-5G
SIAL107840-100GEA
439
EUR
SIAL107840-100G
SIAL107840-25G
SIAL107840-5G
α,α,α-Trifluoroacetophenone, Sigma-Aldrich®
α,α,α-Trifluoroacetophenone
Formula:
C₈H₅F₃O MW: 174.12 g/mol Boiling Pt: 165 Melting Pt: –40 °C Density: 1.2799 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000420 CAS Number: 434-45-7 EINECS: 207-103-1 UN: 1224 ADR: 3 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...