α,α,α-Trifluoro-m-toluic acid
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-(Trifluoromethyl)benzoic acid
DP01925.1
DP01925EA
DP01925EE
MAYBDP01925.1EA
0
EUR
MAYBDP01925.1
MAYBDP01925EA
MAYBDP01925EE
α,α,α-Trifluoro-m-toluic acid
α,α,α-Trifluoro-m-toluic acid
Formula:
C₈H₅F₃O₂ MW: 190.12 g/mol Boiling Pt: 238 Melting Pt: 106 Density: 1.371 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002519 CAS Number: 454-92-2 EINECS: 207-230-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...