α,α,α-Trifluoro-4-iodotoluene, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-Iodobenzotrifluoride, 1-Iodo-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-iodotoluene
M-Clarity™ quality level = MQ100
365858-1G
365858-5G
SIAL365858-1GEA
34.4
EUR
SIAL365858-1G
SIAL365858-5G
α,α,α-Trifluoro-4-iodotoluene, Sigma-Aldrich®
α,α,α-Trifluoro-4-iodotoluene
Formula:
C₇H₄F₃I MW: 272.01 g/mol Boiling Pt: 183…185 °C (1013 hPa) Melting Pt: 17 Density: 1.851 g/cm³ (26 °C) |
MDL Number:
MFCD00039398 CAS Number: 455-13-0 EINECS: 207-234-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...