4-(Trifluoromethyl)phenyl isocyanate
Supplier: Thermo Scientific
Synonyms: α,α,α-Trifluoro-p-tolyl isocyanate, 1-Isocyanato-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)phenylisocyanate, p-Trifluoromethylphenylisocyanate
SB01857.1 SB01857.10
MAYBSB01857.1EA 0 EUR
MAYBSB01857.1 MAYBSB01857.10
4-(Trifluoromethyl)phenyl isocyanate
4-(Trifluoromethyl)phenyl isocyanate
Formula: C₈H₄F₃NO
MW: 187.12 g/mol
Boiling Pt: 173
Melting Pt: 58…59 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002028
CAS Number: 1548-13-6
EINECS: 216-284-6
UN: 2285
ADR: 6.1 II