4-(Trifluoromethyl)phenyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Isocyanato-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-tolyl isothiocyanate, p-Trifluoromethylphenylisothiocyanate
MQ100
476099-1G 476099-5G
SIAL476099-1GEA 53.6 EUR
SIAL476099-1G SIAL476099-5G
4-(Trifluoromethyl)phenyl isothiocyanate, Sigma-Aldrich®
4-(Trifluoromethyl)phenyl isothiocyanate
Formula: C₈H₄F₃NS
MW: 203.19 g/mol
Boiling Pt: 205…207 °C (1013 hPa)
Melting Pt: 29…31 °C
MDL Number: MFCD00039645
CAS Number: 1645-65-4
UN: 2923
ADR: 8 III