3-Fluorobenzylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
(3-Fluorophenyl)methanamine
M-Clarity™ quality level = MQ100
126896-1G
126896-5G
SIAL126896-1GEA
21.3
EUR
SIAL126896-1G
SIAL126896-5G
3-Fluorobenzylamine, Sigma-Aldrich®
3-Fluorobenzylamine
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 87 Density: 1.09 g/cm³ (20 °C) |
MDL Number:
MFCD00008113 CAS Number: 100-82-3 EINECS: 202-891-3 UN: 2735 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...