α,α,α-Trifluoro-o-toluidine
Supplier: Thermo Scientific
Synonyms: 2-(Trifluoromethyl)aniline, 2-Aminobenzotrifluoride
SB00885DA SB00885EA SB00885.50
MAYBSB00885DAEA 0 EUR
MAYBSB00885DA MAYBSB00885EA MAYBSB00885.50
α,α,α-Trifluoro-o-toluidine
α,α,α-Trifluoro-o-toluidine
Formula: C₇H₆F₃N
MW: 161.13 g/mol
Boiling Pt: 175
Melting Pt: –33 °C
Density: 1.282 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007718
CAS Number: 88-17-5
EINECS: 201-806-7
UN: 2942
ADR: 6.1 III