α,α,α-Trifluoro-p-toluidine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Aminobenzotrifluoride, 4-(Trifluoromethyl)aniline, Leflunomide impurity A

M-Clarity™ quality level = MQ200

224936-25G 224936-10G 224936-1G
SIAL224936-25GEA 102 EUR
SIAL224936-25G SIAL224936-10G SIAL224936-1G
α,α,α-Trifluoro-p-toluidine, Sigma-Aldrich®
α,α,α-Trifluoro-p-toluidine
Formula: C₇H₆F₃N
Boiling Pt: 190 °C (1013 hPa)
Melting Pt: 3…8 °C
Density: 1.3 g/cm³ (20 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00064396
CAS Number: 455-14-1
EINECS: 207-236-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR