4-Nitro-2-(trifluoromethyl)aniline
Supplier: Thermo Scientific
Warning
Synonyms: 2-Amino-5-nitro-α,α,α-trifluortoluene, 2-Amino-5-nitro-α,α,α-trifluorotoluene, 4-Nitro-2-trifluoromethylaniline, 2-Amino-5-nitrobenzotrifluoride, 4-Nitro-α,α,α-trifluoro-o-toluidine
TL00267.1 TL00267.10 TL00267.50
MAYBTL00267.1EA 0 EUR
MAYBTL00267.1 MAYBTL00267.10 MAYBTL00267.50
4-Nitro-2-(trifluoromethyl)aniline
4-Nitro-2-(trifluoromethyl)aniline
Formula: C₇H₅F₃N₂O₂
MW: 206.12 g/mol
Melting Pt: 94…95 °C
Density: 1.754 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007365
CAS Number: 121-01-7
EINECS: 204-443-2
UN: 2811
ADR: 6.1 III