α,α,α-Trifluoro-p-cresol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 4-Hydroxybenzotrifluoride, 4-(Trifluoromethyl)phenol

178470-1G 178470-25G 178470-5G
SIAL178470-1GEA 54.4 EUR
SIAL178470-1G SIAL178470-25G SIAL178470-5G
α,α,α-Trifluoro-p-cresol, Sigma-Aldrich®
α,α,α-Trifluoro-p-cresol
Formula: C₇H₅F₃O
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 45…47 °C
Density: 1,3 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002363
CAS Number: 402-45-9
EINECS: 206-945-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR