4'-Iodoacetophenone 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone
278890250 278890050
ACRO278890250EA 115 EUR
ACRO278890250 ACRO278890050
4'-Iodoacetophenone 98%
4'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246.05 g/mol
Melting Pt: 82…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8
Order Now
Specification Test Results
Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 82 to 89 °C
GC ≥97.5 %