2-Ethylphenyl isothiocyanate

Supplier: Thermo Scientific

SB01715.1 SB01715EA SB01715.50
MAYBSB01715.1EA 0 EUR
MAYBSB01715.1 MAYBSB01715EA MAYBSB01715.50
2-Ethylphenyl isothiocyanate
2-Ethylphenyl isothiocyanate
Formula: C₉H₉NS
Boiling Pt: 53 °C (0.1 mmHg)
Density: 1,076 g/cm³ (20 °C)
MDL Number: MFCD00041066
CAS Number: 19241-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR