m-Toluidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 3-Aminotoluene, 3-Toluidine, 3-Methylaniline
M-Clarity™ quality level = MQ200
511218-5ML 511218-100ML 511218-1L
SIAL511218-5MLEA 45.6 EUR
SIAL511218-5ML SIAL511218-100ML SIAL511218-1L
m-Toluidine, Sigma-Aldrich®
m-Toluidine
Formula: C₇H₉N
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.98444 g/cm³ (20 °C)
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007808
CAS Number: 108-44-1
EINECS: 203-583-1
UN: 1708
ADR: 6.1,II
Merck Index: 13,09614