3-Methoxybenzylamine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole
M-Clarity™ quality level = MQ100
159891-1G 159891-5G
SIAL159891-1GEA 51.4 EUR
SIAL159891-1G SIAL159891-5G
3-Methoxybenzylamine, Sigma-Aldrich®
3-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 238
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00008115
CAS Number: 5071-96-5
EINECS: 225-779-6
UN: 2735
ADR: 8 III