2-Aminobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: (2-Aminophenyl)methanol, 2-Hydroxymethylaniline, 2-(Hydroxymethyl)aniline, 2-Aminobenzyl Alcohol

M-Clarity™ quality level = MQ100

122831-10G 122831-25G
SIAL122831-10GEA 41.3 EUR
SIAL122831-10G SIAL122831-25G
2-Aminobenzyl alcohol, Sigma-Aldrich®
2-Aminobenzyl alcohol
Formula: C₇H₉NO
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007749
CAS Number: 5344-90-1
EINECS: 226-293-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR