2-Methyl-4-nitrophenol, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

422908-100G
SIAL422908-100GEA 178 EUR
SIAL422908-100G
2-Methyl-4-nitrophenol, Sigma-Aldrich®
2-Methyl-4-nitrophenol
Formula: C₇H₇NO₃
Boiling Pt: 311 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01734013
CAS Number: 99-53-6
EINECS: 202-763-7
UN: 2446
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR