2-Phenylethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Phenethyl alcohol, Benzylcarbinol, β-Hydroxyethylbenzene, Benzyl carbinol, Benzenethanol
M-Clarity™ quality level = MQ300
W285811-100G-K W285811-1KG-K W285811-25G-K
SIALW285811-100G-KEA 248 EUR
SIALW285811-100G-K SIALW285811-1KG-K SIALW285811-25G-K
2-Phenylethanol, Sigma-Aldrich®
2-Phenylethanol
Formula: C₆H₅CH₂CH₂OH
MW: 122.17 g/mol
Boiling Pt: 432
Melting Pt: 103…105 °C
Density: 1.02 g/cm³ (20 °C)
Flash Pt: 102
Storage Temperature: Ambient
MDL Number: MFCD00002886
CAS Number: 60-12-8
EINECS: 200-456-2
UN: 2810
ADR: 6.1 III
Merck Index: 12,07370