Benzyl alcohol ≥98%, GPR RECTAPUR®

Supplier: VWR Chemicals

20805.291EA 86 EUR
20805.291
Benzyl alcohol ≥98%, GPR RECTAPUR®
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108,14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15,3 °C
Density: 1,05 g/cm³ (20 °C)
Flash Pt: 101 °C
MDL Number: MFCD00004599
CAS Number: 100-51-6
Merck Index: 13,01126
https://ru.vwr.com/stibo/web/std.lang.all/97/87/2299787.jpg

Order Now

Specification Test Results

Assay Min. 98 %
Boiling point 204 - 208 °C
Density (20/4) 1.043 - 1.046

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR