Benzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol
M-Clarity™ quality level = MQ100
305197-100ML 305197-1L 305197-2L
SIAL305197-100MLEA 124 EUR
SIAL305197-100ML SIAL305197-1L SIAL305197-2L
Benzyl alcohol, Sigma-Aldrich®
Benzyl alcohol
Formula: C₆H₅CH₂OH
MW: 108.14 g/mol
Boiling Pt: 205
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM