Benzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W213705-1KG-K W213705-25KG-K
SIALW213705-1KG-KEA 76.4 EUR
SIALW213705-1KG-K SIALW213705-25KG-K
Benzyl alcohol, Sigma-Aldrich®
Benzyl alcohol
Formula: C₆H₅CH₂OH
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: –15.3 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM