4-Octylphenol ≥99%
Supplier: Thermo Fisher Scientific
Danger
H26396.03 H26396.06
H26396.03EA 31.6 EUR
H26396.03 H26396.06
4-Octylphenol ≥99%
4-Octylphenol
Purity: 99% CAS No.: 1806-26-4 Molecular Formula: C14H22O Formula Weight: 206.32 Flash Point: 110°(230°F) Melting Point: 43-45° Boiling Point: 150°/4mm UN No.: 2430 MDL No.: MFCD00036134
Formula: C₁₄H₂₂O
Boiling Pt: 150 °C (4 mmHg)
Melting Pt: 43…45 °C
Density: 0.961
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036134
CAS Number: 1806-26-4
EINECS: 217-302-5
UN: 2430
ADR: 8,III