4-tert-Butylpyrocatechol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol
19670-100G 19670-500G
SIAL19670-100GEA 37.1 EUR
SIAL19670-100G SIAL19670-500G
4-tert-Butylpyrocatechol, Sigma-Aldrich®
4-tert-Butylpyrocatechol
Formula: C₁₀H₁₄O₂
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 53 °C
Density: 1.05 g/cm³ (60 °C)
Flash Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00002201
CAS Number: 98-29-3
EINECS: 202-653-9
UN: 2923
ADR: 8,III