Salicyl alcohol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2-Hydroxyphenyl)methanol, 2-(Hydroxymethyl)phenol, 2-Hydroxybenzyl alcohol, Saligenin
M-Clarity™ quality level = MQ200
166952-100G 166952-25G
SIAL166952-100GEA 102 EUR
SIAL166952-100G SIAL166952-25G
Salicyl alcohol, Sigma-Aldrich®
Salicyl alcohol
Formula: C₇H₈O₂
Boiling Pt: 268 °C (1013 hPa)
Melting Pt: 82 °C
Density: 1.1613 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004617
CAS Number: 90-01-7
EINECS: 201-960-5
Merck Index: 12,08477