4-Ethylthiophenol ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-Ethylbenzenethiol, 4-Ethylbenzene-1-thiol, 1-Ethyl-4-mercaptobenzene
B25026.03
B25026.06
B25026.03EA
31.1
EUR
B25026.03
B25026.06
4-Ethylthiophenol ≥97%
4-Ethylthiophenol
Purity: 97%
CAS No.: 4946-13-8
Molecular Formula: C8H10S
Formula Weight: 138.23
Refractive Index: 1.5645
Odour: Stench
Boiling Point: 119-120°/30mm
Beilstein Reference: 1905734
UN No.: 2810
MDL No.: MFCD00040229
Formula:
C₈H₁₀S Boiling Pt: 119…120 °C (30 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00040229 CAS Number: 4946-13-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...