p-Toluenesulphonyl chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Toluene-4-sulphonyl chloride, Tosyl chloride, 4-Toluenesulphonyl chloride, Toluene-p-sulphonyl chloride
240877-100G 240877-500G 240877-5G
SIAL240877-100GEA 120 EUR
SIAL240877-100G SIAL240877-500G SIAL240877-5G
p-Toluenesulphonyl chloride, Sigma-Aldrich®
p-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 67 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: 128 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00007450
CAS Number: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8,II
Merck Index: 13,09612