Nonylbenzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Phenylnonane

M-Clarity™ quality level = MQ100

331066-100G 331066-25G
SIAL331066-100GEA 420 EUR
SIAL331066-100G SIAL331066-25G
Nonylbenzene, Sigma-Aldrich®
Nonylbenzene
Formula: C₁₅H₂₄
MW: 204.36 g/mol
Boiling Pt: 282
Melting Pt: –24 °C
Density: 0.8584 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009575
CAS Number: 1081-77-2
EINECS: 214-103-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR