4-Bromobenzophenone, Sigma-Aldrich®
Supplier: Merck
Synonyms: (4-Bromophenyl)(phenyl)methanone, p-Bromobenzophenone
M-Clarity™ quality level = MQ100
B58601-100G B58601-25G
SIALB58601-100GEA 69.6 EUR
SIALB58601-100G SIALB58601-25G
4-Bromobenzophenone, Sigma-Aldrich®
4-Bromobenzophenone
Formula: C₁₃H₉BrO
Boiling Pt: 350 °C (1013 hPa)
Melting Pt: 78…82 °C
MDL Number: MFCD00000103
CAS Number: 90-90-4
EINECS: 202-024-9