Bromocresol purple, Sigma-Aldrich®
Supplier: Merck
Warning
114375-5G
SIAL114375-5GEA 48.9 EUR
SIAL114375-5G
Bromocresol purple, Sigma-Aldrich®
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
Melting Pt: 242 °C
Storage Temperature: Ambient
MDL Number: MFCD00011681
CAS Number: 115-40-2
EINECS: 204-087-8
Merck Index: 13,01371