Dichloro(diphenyl)methane, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Dichlorodiphenylmethane, α,α-Dichlorodiphenylmethane
M-Clarity™ quality level = MQ100
D61407-100G
D61407-25G
SIALD61407-100GEA
312
EUR
SIALD61407-100G
SIALD61407-25G
Dichloro(diphenyl)methane, Sigma-Aldrich®
Dichloro(diphenyl)methane
Formula:
(C₆H₅)₂CCl₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 148 °C Density: 1.25 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000811 CAS Number: 2051-90-3 EINECS: 218-134-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...