Diphenylmethane, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

D209317-1KG D209317-25G D209317-500G
SIALD209317-1KGEA 136 EUR
SIALD209317-1KG SIALD209317-25G SIALD209317-500G
Diphenylmethane, Sigma-Aldrich®
Diphenylmethane
Formula: C₁₃H₁₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 22…25 °C
Density: 0.9985 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004781
CAS Number: 101-81-5
EINECS: 202-978-6
UN: 3077
ADR: 9,III
Merck Index: 13,03360

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR