Tetraphenylmethane 96%
Supplier: Thermo Fisher Scientific
A14706.03
A14706.06
A14706.14
A14706.03EA
84.2
EUR
A14706.03
A14706.06
A14706.14
Tetraphenylmethane 96%
Tetraphenylmethane
Purity: 96%
CAS No.: 630-76-2
Molecular Formula: C25H20
Formula Weight: 320.44
Melting Point: 280-285°
Boiling Point: 431°
Solubility: Insoluble in water
Beilstein Reference: 1913557
MDL No.: MFCD00014428
Formula:
C₂₅H₂₀ Boiling Pt: 431 °C Melting Pt: 280…285 °C Density: 1.217 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00014428 CAS Number: 630-76-2 EINECS: 211-144-0 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...