5-Bromo-m-xylene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Bromo-3,5-dimethylbenzene
M-Clarity™ quality level = MQ100
276316-100G 276316-25G
SIAL276316-100GEA 130 EUR
SIAL276316-100G SIAL276316-25G
5-Bromo-m-xylene, Sigma-Aldrich®
5-Bromo-m-xylene
Formula: C₈H₉Br
Boiling Pt: 60…62 °C (5.33 hPa)
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000087
CAS Number: 556-96-7