α,α'-Dichloro-o-xylene, Sigma-Aldrich®
Supplier: Merck
Synonyms: o-Xylylene Dichloride, o-Xylylene chloride, α,α′-Dichloro-o-xylene, α,α'-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, Alpha,Alpha’-Dichloro-o-xylene, 1,2-Bis(chloromethyl)benzene, o-Xylene dichloride
M-Clarity™ quality level = MQ200
123889-100G 123889-25G
SIAL123889-100GEA 163 EUR
SIAL123889-100G SIAL123889-25G
α,α'-Dichloro-o-xylene, Sigma-Aldrich®
α,α'-Dichloro-o-xylene
Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 240
Melting Pt: 54…55 °C
Density: 1.393 g/cm³ (0 °C)
MDL Number: MFCD00000903
CAS Number: 612-12-4
EINECS: 210-291-8
UN: 2928
ADR: 6.1 II