2,6-Diisopropylphenyl isocyanate

Supplier: Thermo Scientific

Synonyms: 2,6-Diisopropylphenylisocyanate

AC31412.1 AC31412.10 AC31412.50
MAYBAC31412.1EA 0 EUR
MAYBAC31412.1 MAYBAC31412.10 MAYBAC31412.50
2,6-Diisopropylphenyl isocyanate
2,6-Diisopropylphenyl isocyanate
Formula: C₁₃H₁₇NO
Boiling Pt: 117 °C (1013 hPa)
Density: 1.2485 g/cm³ (20 °C)
MDL Number: MFCD00008882
CAS Number: 28178-42-9
EINECS: 248-885-4
UN: 3382
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR