2,6-Dimethylphenyl isocyanate ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
2,6-Xylyl isocyanate, 2,6-Dimethylphenylisocyanate, 2-Isocyanato-1,3-dimethylbenzene
L12805.06
L12805.14
L12805.03
L12805.06EA
70.4
EUR
L12805.06
L12805.14
L12805.03
2,6-Dimethylphenyl isocyanate ≥98%
2,6-Dimethylphenyl isocyanate
Purity: 98%
CAS No.: 28556-81-2
Molecular Formula: C9H9NO
Formula Weight: 147.18
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.5356
Boiling Point: 71-72°/3mm
Beilstein Reference: 742593
UN No.: 2206
MDL No.: MFCD00002012
Formula:
C₉H₉NO Boiling Pt: 71…72 °C (3 mmHg) Density: 1.057 Flash Pt: 86 °C (186 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002012 CAS Number: 28556-81-2 UN: 2206 ADR: 6.1,II |