2-Nitro-4,5-xylidine

Supplier: Thermo Scientific

BTB06167.1 BTB06167EA BTB06167.50
MAYBBTB06167.1EA 0 EUR
MAYBBTB06167.1 MAYBBTB06167EA MAYBBTB06167.50
2-Nitro-4,5-xylidine
2-Nitro-4,5-xylidine
Formula: C₈H₁₀N₂O₂
Melting Pt: 143 °C
Density: 1.398 g/cm³ (–123.15 °C)
MDL Number: MFCD00007811
CAS Number: 6972-71-0
EINECS: 230-211-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR