3,4-Dimethylanisole, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
137499-250G 137499-50G
SIAL137499-250GEA 379 EUR
SIAL137499-250G SIAL137499-50G
3,4-Dimethylanisole, Sigma-Aldrich®
3,4-Dimethylanisole
Formula: C₉H₁₂O
Boiling Pt: 200 °C (1013 hPa)
Density: 0.97 g/cm³ (20 °C)
MDL Number: MFCD00008396
CAS Number: 4685-47-6
EINECS: 225-142-2