3,4-Xylenol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W359602-1KG W359602-250G W359602-5KG W359602-SAMPLE
SIALW359602-1KGEA 267 EUR
SIALW359602-1KG SIALW359602-250G SIALW359602-5KG SIALW359602-SAMPLE
3,4-Xylenol, Sigma-Aldrich®
3,4-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 62…65 °C
Density: 1.023 g/cm³ (17 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002304
CAS Number: 95-65-8
EINECS: 202-439-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215