3,5-Xylenol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
144134-100G 144134-5G
SIAL144134-100GEA 45.7 EUR
SIAL144134-100G SIAL144134-5G
3,5-Xylenol, Sigma-Aldrich®
3,5-Xylenol
Formula: C₈H₁₀O
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 61…64 °C
Density: 0.968 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002307
CAS Number: 108-68-9
EINECS: 203-606-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215