4-Methylbenzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4-(Hydroxymethyl)toluole, p-Tolylmethanol, (4-Methylphenyl)methanol
M-Clarity™ quality level = MQ100
127809-5G
127809-100G
127809-25G
SIAL127809-5GEA
28.9
EUR
SIAL127809-5G
SIAL127809-100G
SIAL127809-25G
4-Methylbenzyl alcohol, Sigma-Aldrich®
4-Methylbenzyl alcohol
Formula:
C₈H₁₀O Boiling Pt: 217 °C (1013 hPa) Melting Pt: 59 °C Density: 0.978 g/cm³ (72 °C) |
MDL Number:
MFCD00004664 CAS Number: 589-18-4 EINECS: 209-639-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...