1,4-Diisopropylbenzene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
126276-100G 126276-25G
SIAL126276-100GEA 95.6 EUR
SIAL126276-100G SIAL126276-25G
1,4-Diisopropylbenzene, Sigma-Aldrich®
1,4-Diisopropylbenzene
Formula: C₁₂H₁₈
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: –40 °C
Density: 0.87707 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008892
CAS Number: 100-18-5
EINECS: 202-826-9